siRNA Details
| Gene Symbol: Rab11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017382.5 | NP_059078.2 | 4 | 599 | ||
| GenBank mRNA | AB232605.1 | - | 4 | 466 | ||
| AK014268.1 | - | 4 | 600 | |||
| AK031143.1 | - | 4 | 502 | |||
| AK132072.1 | - | 4 | 583 | |||
| AK132159.1 | - | 4 | 585 | |||
| AK147122.1 | - | 4 | 600 | |||
| AK219017.1 | - | 1 | 43 | |||
| BC010722.1 | - | 4 | 588 | 2811973 | ||
| D50500.1 | - | 2 | 107 | |||

Rab11a
Genomic Map