siRNA Details
| Gene Symbol: Nphp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291012.1 | NP_001277941.1 | 11 | 1045 | ||
| NM_001291013.1 | NP_001277942.1 | 11 | 1042 | |||
| NM_016902.4 | NP_058598.1 | 11 | 1048 | |||
| GenBank mRNA | AF127180.1 | - | 11 | 999 | ||
| AF176210.1 | - | 11 | 1011 | |||
| AJ243223.1 | - | 11 | 996 | |||
| AK159627.1 | - | 11 | 1022 | |||
| BC118953.2 | - | 11 | 993 | 40093235 | ||
| EF174553.3 | - | 11 | 1017 | |||

Nphp1
Genomic Map