siRNA Details
| Gene Symbol: Slc7a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017394.4 | NP_059090.3 | 6 | 966 | ||
| GenBank mRNA | AB026688.1 | - | 6 | 880 | ||
| AK005282.1 | - | 4 | 363 | |||
| AK031867.1 | - | 6 | 934 | |||
| BC054765.1 | - | 6 | 909 | 6414759 | ||

Slc7a10
Genomic Map