siRNA Details
| Gene Symbol: Actr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016860.1 | NP_058556.1 | 5 | 401 | ||
| GenBank mRNA | AB010297.1 | - | 5 | 401 | ||
| AK010632.1 | - | 5 | 398 | |||
| AK019518.1 | - | 5 | 405 | |||
| AK162248.1 | - | 5 | 416 | |||
| AK167450.1 | - | 5 | 408 | |||
| AK168889.1 | - | 5 | 405 | |||
| AK179280.1 | - | 2 | 68 | |||

Actr1a
Genomic Map