siRNA Details
| Gene Symbol: Kat6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205241.1 | NP_001192170.1 | 10 | 1736 | ||
| NM_017479.3 | NP_059507.2 | 10 | 1860 | |||
| GenBank mRNA | AF222800.1 | - | 10 | 1776 | ||
| AK045188.1 | - | 10 | 2199 | |||
| AK147509.1 | - | 10 | 1737 | |||
| BC095974.1 | - | 9 | 1513 | |||

Kat6b
Genomic Map