siRNA Details
| Gene Symbol: Kat6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205241.1 | NP_001192170.1 | 2 | 710 | ||
| NM_017479.3 | NP_059507.2 | 2 | 834 | |||
| GenBank mRNA | AF222800.1 | - | 2 | 750 | ||
| AK045188.1 | - | 2 | 846 | |||
| AK048336.1 | - | 2 | 853 | |||
| AK052307.1 | - | 2 | 766 | |||
| AK083123.1 | - | 2 | 849 | |||
| AK129129.1 | - | 1 | 266 | |||
| AK135465.1 | - | 2 | 886 | |||
| AK147509.1 | - | 2 | 711 | |||
| BC095974.1 | - | 1 | 487 | |||

Kat6b
Genomic Map