siRNA Details
Gene Symbol: Akap8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017476.2 | NP_059504.2 | 4 | 366 | ||
GenBank mRNA | AB028921.1 | - | 4 | 359 | ||
AK011325.1 | - | 4 | 371 | |||
BC010401.1 | - | 4 | 416 | |||
BC046801.1 | - | 4 | 355 | 6403966 | ||
BC051015.1 | - | 4 | 366 |
