siRNA Details
| Gene Symbol: Sult2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017465.2 | NP_059493.2 | 3 | 488 | ||
| GenBank mRNA | AF026072.1 | - | 3 | 455 | ||
| AF478566.1 | - | 2 | 611 | |||
| BC009811.1 | - | 3 | 530 | 4008307 | ||
| BC009813.1 | - | 3 | 418 | 4039050 | ||

Sult2b1
Genomic Map