siRNA Details
| Gene Symbol: Zfp316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017467.3 | NP_059495.3 | 3 | 384 | ||
| GenBank mRNA | BC057078.1 | - | 3 | 301 | 5698016 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp316| Gene Symbol: Zfp316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017467.3 | NP_059495.3 | 3 | 384 | ||
| GenBank mRNA | BC057078.1 | - | 3 | 301 | 5698016 | |