siRNA Details
| Gene Symbol: Cd160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163496.1 | NP_001156968.1 | 4 | 642 | ||
| NM_018767.3 | NP_061237.3 | 4 | 565 | |||
| GenBank mRNA | AF060982.1 | - | 4 | 536 | ||
| AK042093.1 | - | 4 | 566 | |||
| AK139789.1 | - | 4 | 643 | |||
| AK145332.1 | - | 4 | 623 | |||
| BC021596.1 | - | 4 | 537 | 5098004 | ||

Cd160
Genomic Map