siRNA Details
| Gene Symbol: Slc23a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018824.2 | NP_061294.2 | 14 | 1789 | ||
| GenBank mRNA | AB038145.1 | - | 13 | 1525 | ||
| AK122227.1 | - | 10 | 1561 | |||
| AY004874.1 | - | 14 | 1743 | |||
| BC050823.1 | - | 13 | 1553 | 6332451 | ||

Slc23a2
Genomic Map