siRNA Details
| Gene Symbol: Atf7ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019426.2 | NP_062299.2 | 7, 8 | 2301 | ||
| GenBank mRNA | AJ132702.1 | - | 7, 8 | 2294 | ||
| AK132359.1 | - | 6, 7 | 921 | |||
| AK167542.1 | - | 8, 9 | 2347 | |||
| BC037617.1 | - | 2303 | ||||
| BC052460.1 | - | 7, 8 | 2301 | 6834550 | ||
| BC060246.1 | - | 7, 8 | 2284 | 5705945 | ||

Atf7ip
Genomic Map