siRNA Details
| Gene Symbol: Skap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018773.2 | NP_061243.1 | 9 | 856 | ||
| GenBank mRNA | AB014485.1 | - | 9 | 773 | ||
| AF051324.1 | - | 9 | 771 | |||
| AK076000.1 | - | 9 | 801 | |||
| AK144289.1 | - | 9 | 793 | |||
| BC003711.1 | - | 9 | 856 | 3599914 | ||

Skap2
Genomic Map