siRNA Details
| Gene Symbol: Zfp326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018759.2 | NP_061229.2 | 6 | 891 | ||
| GenBank mRNA | AB012725.1 | - | 6 | 857 | ||
| AK017693.1 | - | 6 | 892 | |||
| AK146438.1 | - | 6 | 872 | |||
| BC005567.1 | - | 6 | 882 | |||
| BC037055.1 | - | 4 | 432 | 4948371 | ||
| BC039041.1 | - | 5 | 2896 | |||
| BC042769.1 | - | 1 | 1787 | |||
| BU503985.1 | - | 3 | 275 | |||

Zfp326
Genomic Map