siRNA Details
Gene Symbol: Zfp326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018759.2 | NP_061229.2 | 5, 6 | 730 | ||
GenBank mRNA | AB012725.1 | - | 5, 6 | 696 | ||
AK017693.1 | - | 5, 6 | 731 | |||
AK146438.1 | - | 5, 6 | 711 | |||
BC005567.1 | - | 5, 6 | 721 | |||
BC039041.1 | - | 4, 5 | 2735 |
