siRNA Details
| Gene Symbol: Mtmr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040699.1 | NP_001035789.1 | 11 | 1353 | ||
| GenBank mRNA | AF073882.1 | - | 4 | 412 | ||
| AK028160.1 | - | 11 | 1319 | |||
| AK081973.1 | - | 11 | 1339 | |||
| AK148768.1 | - | 11 | 1354 | |||
| BC032254.1 | - | 11 | 1355 | 5368816 | ||

Mtmr7
Genomic Map