siRNA Details
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019438.1 | NP_062311.1 | 6 | 930 | ||
GenBank mRNA | CX564513.1 | - | 5 | 575 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ncapg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019438.1 | NP_062311.1 | 6 | 930 | ||
GenBank mRNA | CX564513.1 | - | 5 | 575 |