siRNA Details
| Gene Symbol: Crlf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277106.1 | NP_001264035.1 | 4 | 528 | ||
| NM_018776.2 | NP_061246.1 | 4 | 528 | |||
| GenBank mRNA | AF046060.1 | - | 4 | 503 | ||
| AF120152.1 | - | 4 | 437 | |||
| AK037880.1 | - | 4 | 476 | |||
| AK078569.1 | - | 4 | 473 | |||
| AK137528.1 | - | 4 | 480 | |||
| AK146798.1 | - | 4 | 476 | |||
| BC004780.1 | - | 4 | 475 | 3257685 | ||

Crlf3
Genomic Map