siRNA Details
| Gene Symbol: Crlf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018776.2 | NP_061246.1 | 7, 8 | 1147 | ||
| GenBank mRNA | AF046060.1 | - | 7, 8 | 1122 | ||
| AF120152.1 | - | 7, 8 | 1056 | |||
| AK037880.1 | - | 7, 8 | 1095 | |||
| AK137528.1 | - | 7, 8 | 1099 | |||
| AK146798.1 | - | 7, 8 | 1095 | |||
| AK178399.1 | - | 2, 3 | 186 | |||

Crlf3
Genomic Map