siRNA Details
| Gene Symbol: Crlf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277106.1 | NP_001264035.1 | 2 | 289 | ||
| NM_018776.2 | NP_061246.1 | 2 | 289 | |||
| GenBank mRNA | AF046060.1 | - | 2 | 264 | ||
| AF120152.1 | - | 2 | 198 | |||
| AK037880.1 | - | 2 | 237 | |||
| AK078569.1 | - | 2 | 234 | |||
| AK137528.1 | - | 2 | 241 | |||
| AK146798.1 | - | 2 | 237 | |||
| AK165640.1 | - | 2 | 239 | |||
| BC004780.1 | - | 2 | 236 | 3257685 | ||

Crlf3
Genomic Map