siRNA Details
| Gene Symbol: Stk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019442.3 | NP_062315.1 | 3 | 332 | ||
| GenBank mRNA | AF099934.1 | - | 3 | 266 | ||
| AK146739.1 | - | 3 | 334 | |||
| AK168865.1 | - | 3 | 334 | |||
| AK215314.1 | - | 2 | 92 | |||
| AK218509.1 | - | 2 | 161 | |||
| BC022681.1 | - | 3 | 332 | 4240347 | ||

Stk19
Genomic Map