siRNA Details
| Gene Symbol: Stk19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019442.3 | NP_062315.1 | 2 | 154 | ||
| GenBank mRNA | AF099934.1 | - | 2 | 88 | ||
| AK146739.1 | - | 2 | 156 | |||
| AK168865.1 | - | 2 | 156 | |||
| BC022681.1 | - | 2 | 154 | 4240347 | ||

Stk19
Genomic Map