siRNA Details
| Gene Symbol: Asah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018830.1 | NP_061300.1 | 17 | 1928 | ||
| GenBank mRNA | AB037111.1 | - | 18 | 2213 | ||
| AB037181.1 | - | 17 | 1928 | |||
| AK080951.1 | - | 17 | 2105 | |||
| BC022604.1 | - | 8 | 710 | |||
| BC137898.1 | - | 17 | 1911 | 8860917 | ||
| BC145016.1 | - | 17 | 1911 | 9053564 | ||

Asah2
Genomic Map