siRNA Details
| Gene Symbol: Il1f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019450.3 | NP_062323.1 | 3, 4 | 286 | ||
| GenBank mRNA | AF200493.1 | - | 2, 3 | 125 | ||
| AF206697.1 | - | 3, 4 | 200 | |||
| AK004061.1 | - | 3, 4 | 286 | |||
| AK224931.1 | - | 3, 4 | 220 | |||
| AK224932.1 | - | 3, 4 | 233 | |||
| BC117102.1 | - | 2, 3 | 159 | 40126421 | ||
| BC117104.1 | - | 2, 3 | 159 | 40126423 | ||

Il1f6
Genomic Map