siRNA Details
| Gene Symbol: Mefv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161790.1 | NP_001155262.1 | 7, 8 | 2061 | ||
| NM_001161791.1 | NP_001155263.1 | 7, 8 | 1971 | |||
| NM_019453.2 | NP_062326.2 | 7, 8 | 1971 | |||
| GenBank mRNA | AF143409.1 | - | 7, 8 | 1944 | ||
| BC108993.1 | - | 7, 8 | 1932 | 40047838 | ||
| BC108994.1 | - | 7, 8 | 2022 | 40047839 | ||

Mefv
Genomic Map