siRNA Details
| Gene Symbol: Nup210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018815.2 | NP_061285.2 | 33 | 4635 | ||
| GenBank mRNA | AF113751.1 | - | 33 | 4603 | ||
| AK036378.1 | - | 17 | 2233 | |||
| AK090317.1 | - | 11 | 1418 | |||
| AK173056.1 | - | 33 | 4589 | |||
| BC052468.1 | - | 33 | 4496 | 6835038 | ||

Nup210
Genomic Map