siRNA Details
| Gene Symbol: Nup210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018815.2 | NP_061285.2 | 25 | 3365 | ||
| GenBank mRNA | AF113751.1 | - | 25 | 3333 | ||
| AK036378.1 | - | 9 | 962 | |||
| AK090317.1 | - | 3 | 147 | |||
| AK157272.1 | - | 25 | 3357 | |||
| AK173056.1 | - | 25 | 3319 | |||
| BC052468.1 | - | 25 | 3226 | 6835038 | ||

Nup210
Genomic Map