siRNA Details
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018814.3 | NP_061284.2 | 6 | 1239 | ||
GenBank mRNA | CF746564.1 | - | 5 | 662 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018814.3 | NP_061284.2 | 6 | 1239 | ||
GenBank mRNA | CF746564.1 | - | 5 | 662 |