siRNA Details
| Gene Symbol: Ubqln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018798.2 | NP_061268.2 | 1 | 516 | ||
| GenBank mRNA | AF177346.1 | - | 1 | 517 | ||
| BC053022.1 | - | 1 | 516 | 5708544 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubqln2| Gene Symbol: Ubqln2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018798.2 | NP_061268.2 | 1 | 516 | ||
| GenBank mRNA | AF177346.1 | - | 1 | 517 | ||
| BC053022.1 | - | 1 | 516 | 5708544 | ||