siRNA Details
| Gene Symbol: Nphs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019459.2 | NP_062332.2 | 2 | 231 | ||
| GenBank mRNA | AB513652.1 | - | 2 | 103 | ||
| AF168466.1 | - | 2 | 103 | |||
| AF172256.1 | - | 2 | 231 | |||
| AF191090.1 | - | 2 | 559 | |||
| AK141081.1 | - | 2 | 254 | |||

Nphs1
Genomic Map