siRNA Details
| Gene Symbol: Sfmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166531.1 | NP_001160003.1 | 17, 18 | 1991 | ||
| NM_001166532.1 | NP_001160004.1 | 16, 17 | 2085 | |||
| NM_019460.2 | NP_062333.1 | 16, 17 | 1942 | |||
| GenBank mRNA | AB032165.1 | - | 15, 16 | 1787 | ||
| AK142330.1 | - | 16, 17 | 1970 | |||
| AK154826.1 | - | 16, 17 | 2087 | |||
| AK160573.1 | - | 17, 18 | 1991 | |||
| BC018217.1 | - | 16, 17 | 1953 | |||
| BC036972.1 | - | 16, 17 | 2012 | |||
| BC066188.1 | - | 16, 17 | 1895 | 30476111 | ||

Sfmbt1
Genomic Map