siRNA Details
| Gene Symbol: Sfmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166531.1 | NP_001160003.1 | 10 | 1287 | ||
| NM_001166532.1 | NP_001160004.1 | 9 | 1381 | |||
| NM_019460.2 | NP_062333.1 | 9 | 1238 | |||
| GenBank mRNA | AB032165.1 | - | 8 | 1083 | ||
| AK142330.1 | - | 9 | 1265 | |||
| AK154826.1 | - | 9 | 1383 | |||
| AK160573.1 | - | 10 | 1287 | |||
| BC018217.1 | - | 9 | 1249 | |||
| BC036972.1 | - | 9 | 1308 | |||
| BC066188.1 | - | 9 | 1191 | 30476111 | ||

Sfmbt1
Genomic Map