siRNA Details
| Gene Symbol: Sfmbt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166531.1 | NP_001160003.1 | 4 | 323 | ||
| NM_001166532.1 | NP_001160004.1 | 3 | 417 | |||
| NM_019460.2 | NP_062333.1 | 3 | 274 | |||
| GenBank mRNA | AB032165.1 | - | 2 | 119 | ||
| AK142330.1 | - | 3 | 302 | |||
| AK154826.1 | - | 3 | 419 | |||
| AK160573.1 | - | 4 | 323 | |||
| BC018217.1 | - | 3 | 285 | |||
| BC036972.1 | - | 3 | 344 | |||
| BC066188.1 | - | 3 | 227 | 30476111 | ||
| CJ077633.1 | - | 3 | 274 | |||

Sfmbt1
Genomic Map