siRNA Details
| Gene Symbol: Atp8b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001488.3 | NP_001001488.2 | 5, 6 | 813 | ||
| GenBank mRNA | AK036959.1 | - | 5, 6 | 734 | ||
| AK157316.1 | - | 5, 6 | 1081 | |||
| AY506548.1 | - | 5, 6 | 542 | |||
| BC117946.2 | - | 5, 6 | 540 | 40098035 | ||

Atp8b1
Genomic Map