siRNA Details
| Gene Symbol: Adgrg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173036.3 | NP_766624.3 | 11 | 1334 | ||
| GenBank mRNA | AK037149.1 | - | 7 | 2305 | ||
| AK154648.1 | - | 11 | 1310 | |||
| AK155164.1 | - | 11 | 1306 | |||
| BC026426.1 | - | 9 | 1040 | |||
| BC064726.1 | - | 11 | 1294 | 30433392 | ||
| BC071188.1 | - | 9 | 1270 | |||
More Information
Set Membership: |
|

Adgrg3
Genomic Map