siRNA Details
| Gene Symbol: Dfna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018769.3 | NP_061239.1 | 4 | 559 | ||
| GenBank mRNA | AF073309.1 | - | 4 | 560 | ||
| AK016561.1 | - | 927 | ||||
| AK089459.1 | - | 546 | ||||
| AK155135.1 | - | 538 | ||||
| AK171278.1 | - | 4 | 558 | |||
| BC051962.1 | - | 4 | 619 | |||
| BC086618.1 | - | 4 | 619 | |||
| BC132303.1 | - | 3 | 530 | 40130580 | ||

Dfna5
Genomic Map