siRNA Details
| Gene Symbol: Gbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289492.1 | NP_001276421.1 | 12 | 2235 | ||
| NM_001289493.1 | NP_001276422.1 | 11 | 2060 | |||
| NM_018734.3 | NP_061204.3 | 11 | 2097 | |||
| GenBank mRNA | AK132266.1 | - | 11 | 2096 | ||
| BC019195.1 | - | 12 | 2146 | 5036920 | ||
| U44731.1 | - | 11 | 1949 | |||

Gbp3
Genomic Map