siRNA Details
| Gene Symbol: Fnbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018828.2 | NP_061298.1 | 16 | 3137 | ||
| GenBank mRNA | AK013152.1 | - | 2 | 200 | ||
| AK084018.1 | - | 16 | 2929 | |||
| AK129263.1 | - | 15 | 4008 | |||
| U40750.2 | - | 16 | 3137 | |||

Fnbp4
Genomic Map