siRNA Details
| Gene Symbol: Fnbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018828.2 | NP_061298.1 | 15 | 2770 | ||
| GenBank mRNA | AK084018.1 | - | 15 | 2562 | ||
| AK129263.1 | - | 14 | 3641 | |||
| U40750.2 | - | 15 | 2770 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fnbp4| Gene Symbol: Fnbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018828.2 | NP_061298.1 | 15 | 2770 | ||
| GenBank mRNA | AK084018.1 | - | 15 | 2562 | ||
| AK129263.1 | - | 14 | 3641 | |||
| U40750.2 | - | 15 | 2770 | |||