siRNA Details
| Gene Symbol: Eif3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018749.2 | NP_061219.2 | 14 | 1565 | ||
| GenBank mRNA | AB012580.1 | - | 14 | 1506 | ||
| AK045228.1 | - | 14 | 1507 | |||
| AK049767.1 | - | 14 | 1504 | |||
| AK145689.1 | - | 14 | 1520 | |||
| AK168256.1 | - | 14 | 1555 | |||
| AK205598.1 | - | 1 | 46 | |||
| BC089020.1 | - | 14 | 1499 | 30551991 | ||

Eif3d
Genomic Map