siRNA Details
| Gene Symbol: Eif3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018749.2 | NP_061219.2 | 10 | 1044 | ||
| GenBank mRNA | AB012580.1 | - | 10 | 985 | ||
| AK045228.1 | - | 10 | 986 | |||
| AK049767.1 | - | 10 | 983 | |||
| AK145689.1 | - | 10 | 999 | |||
| AK168256.1 | - | 10 | 1034 | |||
| BC089020.1 | - | 10 | 978 | 30551991 | ||

Eif3d
Genomic Map