siRNA Details
| Gene Symbol: Ebag9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019480.4 | NP_062353.3 | 4 | 516 | ||
| GenBank mRNA | AF076524.1 | - | 3 | 199 | ||
| AK004366.1 | - | 3 | 224 | |||
| AK159499.1 | - | 4 | 566 | |||
| AK160149.1 | - | 4 | 685 | |||
| AY009091.1 | - | 4 | 520 | |||
| BC026717.1 | - | 4 | 271 | |||
| BC108397.1 | - | 4 | 516 | 6329278 | ||
| BC119118.1 | - | 4 | 432 | 8733867 | ||
| BC119120.1 | - | 4 | 432 | 8733869 | ||
| BC144740.1 | - | 4 | 466 | 9053288 | ||

Ebag9
Genomic Map