siRNA Details
| Gene Symbol: Slc1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018861.3 | NP_061349.3 | 3 | 828 | ||
| GenBank mRNA | AF246129.1 | - | 3 | 587 | ||
| AK044280.1 | - | 3 | 799 | |||
| AK136900.1 | - | 3 | 828 | |||
| AK139248.1 | - | 3 | 827 | |||
| AK140866.1 | - | 3 | 829 | |||
| AK148075.1 | - | 3 | 829 | |||
| BC043483.1 | - | 3 | 786 | 5003744 | ||
| BC052733.1 | - | 3 | 702 | 6816689 | ||
| U75215.1 | - | 3 | 698 | |||

Slc1a4
Genomic Map