siRNA Details
| Gene Symbol: Camkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018883.2 | NP_061371.2 | 13 | 1322 | ||
| GenBank mRNA | AF117384.1 | - | 13 | 1277 | ||
| AK144483.1 | - | 12 | 1592 | |||
| BC017529.1 | - | 13 | 1287 | 4924656 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Camkk1| Gene Symbol: Camkk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018883.2 | NP_061371.2 | 13 | 1322 | ||
| GenBank mRNA | AF117384.1 | - | 13 | 1277 | ||
| AK144483.1 | - | 12 | 1592 | |||
| BC017529.1 | - | 13 | 1287 | 4924656 | ||