siRNA Details
| Gene Symbol: Cpxm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018867.5 | NP_061355.3 | 4, 5 | 813 | ||
| GenBank mRNA | AF017639.1 | - | 4, 5 | 814 | ||
| AK019509.1 | - | 4, 5 | 812 | |||
| AK038874.1 | - | 4, 5 | 696 | |||
| BC021444.1 | - | 4, 5 | 735 | 5038528 | ||

Cpxm2
Genomic Map