siRNA Details
| Gene Symbol: Ppie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019489.5 | NP_062362.1 | 2 | 72 | ||
| GenBank mRNA | AF182825.1 | - | 2 | 54 | ||
| AK008177.1 | - | 2 | 73 | |||
| AK042165.1 | - | 1 | 1022 | |||
| BC045154.1 | - | 2 | 67 | 6468589 | ||

Ppie
Genomic Map