siRNA Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207678.1 | NP_997561.1 | 11 | 1517 | ||
| GenBank mRNA | AB041605.1 | - | 11 | 1494 | ||
| AK008585.1 | - | 6 | 511 | |||
| AK048244.1 | - | 11 | 3297 | |||
| AY337018.1 | - | 11 | 1500 | |||
| BC003773.1 | - | 4 | 640 | |||
| BC023747.1 | - | 11 | 3285 | |||
| BC083055.1 | - | 4 | 504 | |||
| BC132295.1 | - | 11 | 1509 | 40130572 | ||
| U37351.1 | - | 4 | 523 | |||

Ccnl2
Genomic Map