siRNA Details
| Gene Symbol: Rala | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019491.5 | NP_062364.3 | 4 | 678 | ||
| GenBank mRNA | AF244951.1 | - | 3 | 441 | ||
| AK013881.1 | - | 4 | 667 | |||
| BC031741.1 | - | 4 | 678 | 4501311 | ||
| Z48587.1 | - | 3 | 441 | |||

Rala
Genomic Map