siRNA Details
| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139520.1 | NP_001132992.1 | 14 | 1716 | ||
| NM_018851.3 | NP_061339.3 | 14 | 1716 | |||
| GenBank mRNA | AK054490.1 | - | 14 | 1615 | ||
| AK078936.1 | - | 14 | 1659 | |||
| AK128954.1 | - | 13 | 1514 | |||
| AK151335.1 | - | 14 | 1590 | |||
| AK153390.1 | - | 14 | 1590 | |||
| AK170688.1 | - | 14 | 1615 | |||
| AK171565.1 | - | 14 | 1616 | |||
| BC012721.1 | - | 14 | 1653 | 4037046 | ||
| BC067198.1 | - | 14 | 1639 | 6826682 | ||
| U15635.1 | - | 14 | 1640 | |||

Samhd1
Genomic Map