siRNA Details
| Gene Symbol: Samhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001139520.1 | NP_001132992.1 | 8, 9 | 1096 | ||
| NM_018851.3 | NP_061339.3 | 8, 9 | 1096 | |||
| GenBank mRNA | AK054490.1 | - | 8, 9 | 995 | ||
| AK078936.1 | - | 8, 9 | 1039 | |||
| AK128954.1 | - | 7, 8 | 894 | |||
| AK149865.1 | - | 8, 9 | 971 | |||
| AK151335.1 | - | 8, 9 | 970 | |||
| AK153390.1 | - | 8, 9 | 970 | |||
| AK170688.1 | - | 8, 9 | 995 | |||
| AK171565.1 | - | 8, 9 | 996 | |||
| BC012721.1 | - | 8, 9 | 1033 | 4037046 | ||
| BC067198.1 | - | 8, 9 | 1019 | 6826682 | ||
| U15635.1 | - | 8, 9 | 1020 | |||

Samhd1
Genomic Map